Learn More
Biomatik Corporation Human Aldehyde Dehydrogenase 1 Family, Member A2 (ALDH1A2) ELISA Kit
Supplier:  Biomatik Corporation EKU02224
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.625-40ng/mL. Sensitivity: 0.223ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: O94788. Target Synonyms: RALDH(II); RALDH2; RALDH2-T; Retinaldehyde Dehydrogenase 2; Retinal dehydrogenase 2; Retinaldehyde-specific dehydrogenase type 2. Restriction: For Research Use Only.
Catalog No. 50-228-5772