Learn More
Biomatik Corporation Rat Amyloid Beta Peptide 1-40 (Ab1-40) ELISA Kit
Supplier:  Biomatik Corporation EKU02329
Encompass_Preferred
Rat Amyloid Beta Peptide 1-40 (Ab1-40) ELISA Kit for detecting Ab1-40 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.84pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P08592. Gene ID: 54226. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5787