Learn More
Biomatik Corporation Mouse Angiopoietin Like Protein 7 (ANGPTL7) ELISA Kit
Supplier:  Biomatik Corporation EKU02379
Encompass_Preferred
Mouse Angiopoietin Like Protein 7 (ANGPTL7) ELISA Kit for detecting ANGPTL7 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8R1Q3. Gene ID: 654812. Target Synonyms: CDT6; AngX; Cornea-Derived Transcript 6; Angiopoietin-related protein 7; Angiopoietin-like factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5798