Learn More
Biomatik Corporation Mouse Insulin Like Growth Factor Binding Protein 4 (IGFBP4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04999
Encompass_Preferred
Mouse Insulin Like Growth Factor Binding Protein 4 (IGFBP4) ELISA Kit for detecting IGFBP4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.064ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P47879. Gene ID: 16010. Target Synonyms: BP4; HT29-IGFBP; IBP4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2822