Learn More
Biomatik Corporation Human ATP Binding Cassette Transporter A1 (ABCA1) ELISA Kit
Supplier:  Biomatik Corporation EKU02604
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.057ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: O95477. Target Synonyms: CERP; ABC-A1; ABC1; HDLDT1; TGD; Cholesterol Efflux Regulatory Protein; Tangier Disease. Restriction: For Research Use Only.
Catalog No. 50-228-5826