Learn More
Biomatik Corporation Rat Insulin Like Growth Factor Binding Protein 5 (IGFBP5) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05004
Encompass_Preferred
Rat Insulin Like Growth Factor Binding Protein 5 (IGFBP5) ELISA Kit for detecting IGFBP5 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.62-1000pg/ml. Sensitivity: 7.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P24594. Gene ID: 25285. Target Synonyms: IBP5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2825