Learn More
Biomatik Corporation Human Insulin Like Growth Factor Binding Protein, Acid Labile Subunit (IGFALS) ELISA Kit, 96 tests
Catalog No. 501502829
Encompass_Preferred
The Human Insulin Like Growth Factor Binding Protein, Acid Labile Subunit (IGFALS) ELISA Kit allows for quantitative determination insulin Like Growth Factor Binding Protein, Acid Labile Subunit in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.66 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P35858. Cross Activity: No significant cross-reactivity or interference between Insulin Like Growth Factor Binding Protein, Acid Labile Subunit and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2829 Supplier Biomatik Corporation Supplier No. EKU05011
The Human Insulin Like Growth Factor Binding Protein, Acid Labile Subunit (IGFALS) ELISA Kit allows for quantitative determination insulin Like Growth Factor Binding Protein, Acid Labile Subunit in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.66 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P35858. Cross Activity: No significant cross-reactivity or interference between Insulin Like Growth Factor Binding Protein, Acid Labile Subunit and analogues was observed. Restriction: For Research Use Only.