Learn More
Biomatik Corporation Mouse Chemokine C-X-C-Motif Receptor 3 (CXCR3) ELISA Kit
Supplier:  Biomatik Corporation EKU03117
Encompass_Preferred
Mouse Chemokine C-X-C-Motif Receptor 3 (CXCR3) ELISA Kit for detecting CXCR3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 32pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O88410. Gene ID: 12766. Target Synonyms: CD183; CKR-L2; CMKAR3; GPR9; IP10r; IP10-R; Mig-R; MigR; G Protein-Coupled Receptor 9; Interferon-inducible protein 10 receptor; IP-10 receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5879