Learn More
Biomatik Corporation Human Claudin 1 (CLDN1) ELISA Kit
Supplier:  Biomatik Corporation EKU03202
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.061ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: O95832. Target Synonyms: CLD1; ILVASC; SEMP1; Senescence-Associated Epithelial Membrane Protein 1. Restriction: For Research Use Only.
Catalog No. 50-228-5891