Learn More
Biomatik Corporation Mouse Insulin Like Protein 3 (INSL3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05013
Encompass_Preferred
Mouse Insulin Like Protein 3 (INSL3) ELISA Kit for detecting INSL3 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O09107. Target Synonyms: RLNL; RLF; Relaxin-Like Factor,Leydig Cell; Leydig insulin-like peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2831