Learn More
Biomatik Corporation Mouse Collagen Type III (COL3) ELISA Kit
Supplier:  Biomatik Corporation EKU03316
Encompass_Preferred
Mouse Collagen Type III (COL3) ELISA Kit for detecting COL3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 2.4-150ng/ml. Sensitivity: 1.1ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08121. Gene ID: 12825. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5903