Learn More
Biomatik Corporation Mouse Insulin Like Protein 5 (INSL5) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05016
Encompass_Preferred
Mouse Insulin Like Protein 5 (INSL5) ELISA Kit for detecting INSL5 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.62-1000pg/ml. Sensitivity: 6.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9WUG6. Gene ID: 23919. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2833