Learn More
Biomatik Corporation Mouse Collagen Type VI (COL6) ELISA Kit
Supplier:  Biomatik Corporation EKU03333
Encompass_Preferred
Mouse Collagen Type VI (COL6) ELISA Kit for detecting COL6 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.17ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8CII2. Gene ID: 98828. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5908