Learn More
Biomatik Corporation Mouse Insulin Promoter Factor 1 (IPF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05018
Encompass_Preferred
Mouse Insulin Promoter Factor 1 (IPF) ELISA Kit for detecting IPF in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P52946. Gene ID: 18609. Target Synonyms: IDX1; STF1; MODY4; PDX1; GSF; IUF1; Pancreatic And Duodenal Homeobox 1; Somatostatin Transcription Factor 1; Glucose-sensitive factor; Insulin upstream factor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2834