Learn More
Biomatik Corporation Mouse Cytokine Receptor Like Factor 1 (CRLF1) ELISA Kit
Supplier:  Biomatik Corporation EKU03618
Encompass_Preferred
Mouse Cytokine Receptor Like Factor 1 (CRLF1) ELISA Kit for detecting CRLF1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9JM58. Gene ID: 12931. Target Synonyms: CISS; CISS1; CLF; CLF-1; NR6; Cytokine-like factor 1; ZcytoR5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5945