Learn More
Biomatik Corporation Rat Elastin (ELN) ELISA Kit
Supplier:  Biomatik Corporation EKU03854
Encompass_Preferred
Rat Elastin (ELN) ELISA Kit for detecting ELN in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.2-2000pg/ml. Sensitivity: 13.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q99372. Gene ID: 25043. Target Synonyms: WBS, WS, SVAS; Tropoelastin; Supravalvular Aortic Stenosis; Williams-Beuren Syndrome. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5976