Learn More
Biomatik Corporation Mouse Integrin Beta 3 (ITGb3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05044
Encompass_Preferred
Mouse Integrin Beta 3 (ITGb3) ELISA Kit for detecting ITGb3 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2000pg/ml. Sensitivity: 12.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O54890. Gene ID: 16416. Target Synonyms: CD61; ITG-B3; GP3A; GP3-A; GpIIIa; Platelet Membrane Glycoprotein IIIa. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2840