Learn More
Biomatik Corporation Human Epigen (EPG) ELISA Kit
Supplier:  Biomatik Corporation EKU03942
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.115ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: EPGN; Epithelial Mitogen Homolog. Restriction: For Research Use Only.
Catalog No. 50-228-5986