Learn More
Biomatik Corporation Human Integrin Beta 5 (ITGb5) ELISA Kit, 96 tests
Catalog No. 501502841
Encompass_Preferred
The Human Integrin Beta 5 (ITGb5) ELISA Kit allows for quantitative determination integrin Beta 5 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.056 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P18084. Cross Activity: No significant cross-reactivity or interference between Integrin Beta 5 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2841 Supplier Biomatik Corporation Supplier No. EKU05046
The Human Integrin Beta 5 (ITGb5) ELISA Kit allows for quantitative determination integrin Beta 5 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.056 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P18084. Cross Activity: No significant cross-reactivity or interference between Integrin Beta 5 and analogues was observed. Restriction: For Research Use Only.