Learn More
Biomatik Corporation Mouse Epithelial Neutrophil Activating Peptide 78 (ENA78) ELISA Kit
Supplier:  Biomatik Corporation EKU03952
Encompass_Preferred
Mouse Epithelial Neutrophil Activating Peptide 78 (ENA78) ELISA Kit for detecting ENA78 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4,000pg/ml. Sensitivity: 27.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50228. Gene ID: 20311. Target Synonyms: CXCL5; SCYB5; Chemokine C-X-C-Motif Ligand 5; Small Inducible Cytokine Subfamily B (Cys-X-Cys) ,Member 5; Neutrophil-activating peptide ENA-78. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5988