Learn More
Biomatik Corporation Human Integrin Beta 6 (ITGb6) ELISA Kit, 96 tests
Catalog No. 501502842
Encompass_Preferred
The Human Integrin Beta 6 (ITGb6) ELISA Kit allows for quantitative determination integrin Beta 6 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.059 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P18564. Cross Activity: No significant cross-reactivity or interference between Integrin Beta 6 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2842 Supplier Biomatik Corporation Supplier No. EKU05047
The Human Integrin Beta 6 (ITGb6) ELISA Kit allows for quantitative determination integrin Beta 6 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.059 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P18564. Cross Activity: No significant cross-reactivity or interference between Integrin Beta 6 and analogues was observed. Restriction: For Research Use Only.