Learn More
Biomatik Corporation Rat Ferritin (FE) ELISA Kit
Supplier:  Biomatik Corporation EKU04068
Encompass_Preferred
Rat Ferritin (FE) ELISA Kit for detecting FE in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.17ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D3ZUG8. Gene ID: 291470. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5998