Learn More
Biomatik Corporation Human Intelectin 1 (ITLN1) ELISA Kit, 96 tests
Catalog No. 501502843
Encompass_Preferred
The Human Intelectin 1 (ITLN1) ELISA Kit allows for quantitative determination intelectin 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 6.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q8WWA0. Cross Activity: No significant cross-reactivity or interference between Intelectin 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2843 Supplier Biomatik Corporation Supplier No. EKU05048
The Human Intelectin 1 (ITLN1) ELISA Kit allows for quantitative determination intelectin 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 6.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q8WWA0. Cross Activity: No significant cross-reactivity or interference between Intelectin 1 and analogues was observed. Restriction: For Research Use Only.