Learn More
Biomatik Corporation Mouse Intelectin 1 (ITLN1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05049
Encompass_Preferred
Mouse Intelectin 1 (ITLN1) ELISA Kit for detecting ITLN1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.5-800pg/ml. Sensitivity: 6.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O88310. Gene ID: 16429. Target Synonyms: HL1; INTL; ITLN; LFR; HIntL; Omentin; Intelectin 1,Galactofuranose Binding; Intestinal Lactoferrin Receptor;Endothelial lectin HL-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2844