Learn More
Biomatik Corporation Human Inter Alpha-Globulin Inhibitor H2 (ITIH2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05053
Encompass_Preferred
The Human Inter Alpha-Globulin Inhibitor H2 (ITIH2) ELISA Kit allows for quantitative determination inter Alpha-Globulin Inhibitor H2 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.42 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P19823. Cross Activity: No significant cross-reactivity or interference between Inter Alpha-Globulin Inhibitor H2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2846