Learn More
Biomatik Corporation Rat Fibroblast Activation Protein Alpha (FAPa) ELISA Kit
Supplier:  Biomatik Corporation EKU04119
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.312-20ng/mL. Sensitivity: 0.122ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: APCE; SIMP; DPPIV; Seprase; 170 kDa melanoma membrane-bound gelatinase; Post-proline cleaving enzyme; Serine integral membrane protease; Surface-expressed protease. Restriction: For Research Use Only.
Catalog No. 50-228-6008