Learn More
Biomatik Corporation Human Inter Alpha-Globulin Inhibitor H3 (ITIH3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05054
Encompass_Preferred
The Human Inter Alpha-Globulin Inhibitor H3 (ITIH3) ELISA Kit allows for quantitative determination inter Alpha-Globulin Inhibitor H3 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 6.25-400 ng/mL. Sensitivity: 2.39 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q06033. Cross Activity: No significant cross-reactivity or interference between Inter Alpha-Globulin Inhibitor H3 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2847