Learn More
Biomatik Corporation Mouse Fibroblast Growth Factor 8, Androgen Induced (FGF8) ELISA Kit
Supplier:  Biomatik Corporation EKU04182
Encompass_Preferred
Mouse Fibroblast Growth Factor 8, Androgen Induced (FGF8) ELISA Kit for detecting FGF8 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P37237. Gene ID: 14179. Target Synonyms: AIGF; HBGF8; Androgen-induced growth factor; Heparin-binding growth factor 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6010