Learn More
Biomatik Corporation Mouse Inter Alpha-Globulin Inhibitor H4 (ITIH4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05056
Encompass_Preferred
Mouse Inter Alpha-Globulin Inhibitor H4 (ITIH4) ELISA Kit for detecting ITIH4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500ng/ml. Sensitivity: 3.3ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: A6X935. Gene ID: 16427. Target Synonyms: IHRP; PK120; H4P; ITIHL1; Plasma Kallikrein-Sensitive Glycoprotein; Plasma kallikrein sensitive glycoprotein 120; Inter-alpha-trypsin inhibitor family heavy chain-related protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2849