Learn More
Biomatik Corporation Human Inter Alpha-Globulin Inhibitor H5 (ITIH5) ELISA Kit, 96 tests
Catalog No. 501502850
Encompass_Preferred
The Human Inter Alpha-Globulin Inhibitor H5 (ITIH5) ELISA Kit allows for quantitative determination inter Alpha-Globulin Inhibitor H5 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.37 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q86UX2. Cross Activity: No significant cross-reactivity or interference between Inter Alpha-Globulin Inhibitor H5 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2850 Supplier Biomatik Corporation Supplier No. EKU05057
The Human Inter Alpha-Globulin Inhibitor H5 (ITIH5) ELISA Kit allows for quantitative determination inter Alpha-Globulin Inhibitor H5 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.37 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q86UX2. Cross Activity: No significant cross-reactivity or interference between Inter Alpha-Globulin Inhibitor H5 and analogues was observed. Restriction: For Research Use Only.