Learn More
Biomatik Corporation Free Thyroxine (fT4) ELISA Kit
Supplier:  Biomatik Corporation EKU04274
Encompass_Preferred
Free Thyroxine (fT4) ELISA Kit for detecting fT4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 1.11-90pg/ml. Sensitivity: 0.42pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: FTI. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6020