Learn More
Biomatik Corporation Dog Intercellular Adhesion Molecule 1 (ICAM1) ELISA Kit, 96 tests
Catalog No. 501502851
Encompass_Preferred
The Dog Intercellular Adhesion Molecule 1 (ICAM1) ELISA Kit allows for quantitative determination intercellular Adhesion Molecule 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.64 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: F1PB95. Cross Activity: No significant cross-reactivity or interference between Intercellular Adhesion Molecule 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2851 Supplier Biomatik Corporation Supplier No. EKU05059
The Dog Intercellular Adhesion Molecule 1 (ICAM1) ELISA Kit allows for quantitative determination intercellular Adhesion Molecule 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.64 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: F1PB95. Cross Activity: No significant cross-reactivity or interference between Intercellular Adhesion Molecule 1 and analogues was observed. Restriction: For Research Use Only.