Learn More
Biomatik Corporation Mouse Gelsolin (GSN) ELISA Kit
Supplier:  Biomatik Corporation EKU04358
Encompass_Preferred
Mouse Gelsolin (GSN) ELISA Kit for detecting GSN in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 37pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P13020. Gene ID: 227753. Target Synonyms: GS; AGEL; ADF; Brevin; Gelsolin Amyloidosis,Finnish Type; Actin-depolymerizing factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6029