Learn More
Biomatik Corporation Mouse Intercellular Adhesion Molecule 1 (ICAM1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05061
Encompass_Preferred
Mouse Intercellular Adhesion Molecule 1 (ICAM1) ELISA Kit for detecting ICAM1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.119ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P13597. Gene ID: 15894. Target Synonyms: CD54; BB2; P3.58; Human Rhinovirus Receptor; Major group rhinovirus receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2853