Learn More
Biomatik Corporation Mouse Glutamate Receptor, Ionotropic, AMPA 2 (GRIA2) ELISA Kit
Supplier:  Biomatik Corporation EKU04449
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.057ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: GLUR2; GLURB; HBGR2; GluR-K2; AMPA-selective glutamate receptor 2. Restriction: For Research Use Only.
Catalog No. 50-228-6038