Learn More
Biomatik Corporation Mouse Intercellular Adhesion Molecule 5 (ICAM5) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05068
Encompass_Preferred
Mouse Intercellular Adhesion Molecule 5 (ICAM5) ELISA Kit for detecting ICAM5 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.62ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q60625. Gene ID: 15898. Target Synonyms: TLCN; TLN; Telencephalin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2856