Learn More
Biomatik Corporation Dog Interferon Alpha (IFNa) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05070
Encompass_Preferred
The Dog Interferon Alpha (IFNa) ELISA Kit allows for quantitative determination interferon Alpha in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 5.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P81255. Cross Activity: No significant cross-reactivity or interference between Interferon Alpha and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2857