Learn More
Biomatik Corporation Human Glycophosphatidylinositol (GPI) ELISA Kit
Supplier:  Biomatik Corporation EKU04525
Encompass_Preferred
Human Glycophosphatidylinositol (GPI) ELISA Kit for detecting GPI in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 246.9-20,000pg/ml. Sensitivity: 92.2pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: GPI Anchor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6045