Learn More
Biomatik Corporation Human Glycosylphosphatidylinositol Specific Phospholipase D1 (GPLD1) ELISA Kit
Supplier:  Biomatik Corporation EKU04537
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.78-50ng/mL. Sensitivity: 0.30ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P80108. Target Synonyms: GPIPLD; GPIPLDM; PIGPLD; PIGPLD1; Glycoprotein phospholipase D; Glycosyl-phosphatidylinositol-specific phospholipase D. Restriction: For Research Use Only.
Catalog No. 50-228-6048