Learn More
Biomatik Corporation Human Interferon Alpha (IFNa) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05074
Encompass_Preferred
The Human Interferon Alpha (IFNa) ELISA Kit allows for quantitative determination interferon Alpha in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 7.8-500 pg/mL. Sensitivity: 3.0 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01562. Cross Activity: No significant cross-reactivity or interference between Interferon Alpha and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2860