Learn More
Biomatik Corporation Human Growth Differentiation Factor 1 (GDF1) ELISA Kit
Supplier:  Biomatik Corporation EKU04582
Encompass_Preferred
Human Growth Differentiation Factor 1 (GDF1) ELISA Kit for detecting GDF1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P27539. Gene ID: 2657. Target Synonyms: Embryonic growth/differentiation factor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6052