Learn More
Biomatik Corporation Pig Interferon Alpha (IFNa) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05077
Encompass_Preferred
Pig Interferon Alpha (IFNa) ELISA Kit for detecting IFNa in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P49879. Target Synonyms: IFNA1; IFL; LeIF D; IFN; IFN-Alpha; IFNA13; IFN-A; IFNAP22; IFN-Alpha 1b; Interferon Alpha 1b; Interferon, Leukocytic; IFN, Leukocyte; Interferon alpha-D. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2862