Learn More
Biomatik Corporation Mouse Growth Differentiation Factor 3 (GDF3) ELISA Kit
Supplier:  Biomatik Corporation EKU04595
Encompass_Preferred
Mouse Growth Differentiation Factor 3 (GDF3) ELISA Kit for detecting GDF3 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 2.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q07104. Gene ID: 14562. Target Synonyms: Vgr2; Vg-Related Gene 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6056