Learn More
Biomatik Corporation Human Interferon Alpha 2 (IFNa2) ELISA Kit, 96 tests
Catalog No. 501502864
Encompass_Preferred
The Human Interferon Alpha 2 (IFNa2) ELISA Kit allows for quantitative determination interferon Alpha 2 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01563. Cross Activity: No significant cross-reactivity or interference between Interferon Alpha 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2864 Supplier Biomatik Corporation Supplier No. EKU05083
The Human Interferon Alpha 2 (IFNa2) ELISA Kit allows for quantitative determination interferon Alpha 2 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01563. Cross Activity: No significant cross-reactivity or interference between Interferon Alpha 2 and analogues was observed. Restriction: For Research Use Only.