Learn More
Biomatik Corporation Mouse Heparanase (HPSE) ELISA Kit
Supplier:  Biomatik Corporation EKU04738
Encompass_Preferred
Mouse Heparanase (HPSE) ELISA Kit for detecting HPSE in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6YGZ1. Gene ID: 15442. Target Synonyms: HPA; HPR1; HPSE1; HSE1; Endo-glucoronidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6065