Learn More
Biomatik Corporation Human Immunoglobulin A2 (IgA2) ELISA Kit
Supplier:  Biomatik Corporation EKU04883
Encompass_Preferred
Human Immunoglobulin A2 (IgA2) ELISA Kit for detecting IgA2 in serum, plasma, saliva, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.94-60ng/ml. Sensitivity: 0.43ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01877. Target Synonyms: IGHA2; Immunoglobulin Heavy Constant Alpha 2; A2m Marker. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6084