Learn More
Biomatik Corporation Pig Interferon Beta (IFNb) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05093
Encompass_Preferred
Pig Interferon Beta (IFNb) ELISA Kit for detecting IFNb in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q29412. Target Synonyms: IFNB1; IFN-B; IFB; IFF; IFNB; Interferon Beta 1 Fibroblast. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2870