Learn More
Biomatik Corporation Chicken Immunoglobulin G (IgG) ELISA Kit
Supplier:  Biomatik Corporation EKU04893
Encompass_Preferred
Chicken Immunoglobulin G (IgG) ELISA Kit for detecting IgG in serum, plasma, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 246.9-20,000ng/ml. Sensitivity: 95.1ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6087