Learn More
Biomatik Corporation Rat Immunoglobulin G (IgG) ELISA Kit
Supplier:  Biomatik Corporation EKU04898
Encompass_Preferred
Rat Immunoglobulin G (IgG) ELISA Kit for detecting IgG in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.23-100ug/ml. Sensitivity: 0.49ug/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6089