Learn More
Biomatik Corporation Indole 3 Acetic Acid (IAA) ELISA Kit
Supplier:  Biomatik Corporation EKU04915
Encompass_Preferred
Sample Type: Tissue or cell culture supernates. Species Reactivity: General species. Detection Range: 2.47-200ng/mL. Sensitivity: 0.88ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Competitive. Assay Time: 2 hours. UniProt ID: Not Available. Target Synonyms: IA-A; Indolylacetic Acid; Indoleacetic Acid; Heteroauxin. Restriction: For Research Use Only.
Catalog No. 50-228-6094